ENA371463403 Enamine

N-(3-chloropropyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD08714917
  • Cas Number : 67680-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109079
Availability details for
Quantity
  • Availability
  • Add to Cart