ENA371463404 Enamine

3-(6-amino-2-methylpyridin-3-yl)prop-2-enoic acid

  • MDL number MFCD20658864
  • Cas Number : 1807920-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253...255
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109102
Availability details for
Quantity
  • Availability
  • Add to Cart