ENA371463408 Enamine

4-(benzylsulfanyl)-2-chloropyrimidine

  • MDL number MFCD16662239
  • Cas Number : 1249555-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109108
Availability details for
Quantity
  • Availability
  • Add to Cart