ENA371463412 Enamine

methyl 2-{[4-(dimethylamino)phenyl]amino}propanoate

  • MDL number MFCD16088601

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109121
Availability details for
Quantity
  • Availability
  • Add to Cart