ENA371463428 Enamine

2-(2,5-dimethylphenyl)pyrrolidine

  • MDL number MFCD05189463
  • Cas Number : 367280-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109148
Availability details for
Quantity
  • Availability
  • Add to Cart