ENA371463438 Enamine

1-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-one

  • MDL number MFCD00833694
  • Cas Number : 2940-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109292
Availability details for
Quantity
  • Availability
  • Add to Cart