ENA371463446 Enamine

3-cyano-4-fluoro-N-(2,2,2-trifluoroethyl)benzene-1-sulfonamide

  • MDL number MFCD18558064
  • Cas Number : 1394040-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109305
Availability details for
Quantity
  • Availability
  • Add to Cart