ENA371463449 Enamine

ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD03407326
  • Cas Number : 368869-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109308

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109308
Availability details for
Quantity
  • Availability
  • Add to Cart