ENA371463452 Enamine

8-[(4-iodophenoxy)methyl]quinoline

  • MDL number MFCD13202772
  • Cas Number : 1300068-28-3
  • Molecular Weight : 361.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109311
Availability details for
Quantity
  • Availability
  • Add to Cart