ENA371463456 Enamine

4H,6H,7H-thieno[3,2-c]thiopyran-2-carboxylic acid

  • MDL number MFCD16045906
  • Cas Number : 933709-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107710
Availability details for
Quantity
  • Availability
  • Add to Cart