ENA371463459 Enamine

2-(N-methyl2-aminobenzamido)benzoic acid

  • MDL number MFCD00597921
  • Cas Number : 83432-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109354
Availability details for
Quantity
  • Availability
  • Add to Cart