ENA371463461 Enamine

methyl 2-chloro-2-(3,5-dichlorophenyl)acetate

  • MDL number MFCD16694253
  • Cas Number : 1250898-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109358
Availability details for
Quantity
  • Availability
  • Add to Cart