ENA371463463 Enamine

1-(4-hydroxy-2,5-dimethoxyphenyl)ethan-1-one

  • MDL number MFCD22378672
  • Cas Number : 13909-71-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-hydroxy-2,5-dimethoxyphenyl)ethanone
Inchi Code 1S/C10H12O4/c1-6(11)7-4-10(14-3)8(12)5-9(7)13-2/h4-5,12H,1-3H3
Inchi Key PKGDSHOPNITYCE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109360
Availability details for
Quantity
  • Availability
  • Add to Cart