ENA371463467 Enamine

trichloropyrimidine-5-carboxylic acid

  • MDL number MFCD09909820
  • Cas Number : 93416-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-153
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107738
Availability details for
Quantity
  • Availability
  • Add to Cart