ENA371463468 Enamine

1-tert-butyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11207433
  • Cas Number : 1094377-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109370
Availability details for
Quantity
  • Availability
  • Add to Cart