ENA371463469 Enamine

3-cyclobutyl-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD11209669
  • Cas Number : 92406-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109371
Availability details for
Quantity
  • Availability
  • Add to Cart