ENA371463480 Enamine

2-methoxy-1-methyl-4-(propan-2-yl)benzene

  • MDL number MFCD00216689
  • Cas Number : 6379-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109392
Availability details for
Quantity
  • Availability
  • Add to Cart