ENA371463483 Enamine

1-(2,4-dichloropyridin-3-yl)ethan-1-one

  • MDL number MFCD11847304
  • Cas Number : 1246349-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109395
Availability details for
Quantity
  • Availability
  • Add to Cart