ENA371463484 Enamine

N-tert-butyl-3-cyano-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD19126022
  • Cas Number : 1275247-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109396

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109396
Availability details for
Quantity
  • Availability
  • Add to Cart