ENA371463493 Enamine

1-(2-ethoxyethyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD16855637
  • Cas Number : 1309166-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109443
Availability details for
Quantity
  • Availability
  • Add to Cart