ENA371463494 Enamine

3-chloro-4-[2-(1H-imidazol-1-yl)ethoxy]benzaldehyde

  • MDL number MFCD16501074
  • Cas Number : 1284907-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109444
Availability details for
Quantity
  • Availability
  • Add to Cart