ENA371463495 Enamine

1-(cyclobutylmethyl)-5-oxopyrrolidine-3-carbonitrile

  • MDL number MFCD19677383
  • Cas Number : 1340167-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109445
Availability details for
Quantity
  • Availability
  • Add to Cart