ENA371463500 Enamine

1-(cyclopropylmethyl)-1H-pyrazol-5-amine

  • MDL number MFCD08457366
  • Cas Number : 660853-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109450
Availability details for
Quantity
  • Availability
  • Add to Cart