ENA371463503 Enamine

1-nitro-4-(2-nitrobut-1-en-1-yl)benzene

  • Cas Number : 1212-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109464
Availability details for
Quantity
  • Availability
  • Add to Cart