ENA371463506 Enamine

4-[2-amino-4-(trifluoromethyl)phenyl]-1$l^{6},4-thiomorpholine-1,1-dione

  • MDL number MFCD12569406
  • Cas Number : 861881-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109474

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109474
Availability details for
Quantity
  • Availability
  • Add to Cart