ENA371463508 Enamine

1-(2-methoxyethyl)cyclopentane-1-carbonitrile

  • MDL number MFCD21002233
  • Cas Number : 64871-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109476

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P311,P312,P330,P361,P363,P403+P233,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-109476
pictograms GHS06
hstatements H301,H311,H331
Availability details for
Quantity
  • Availability
  • Add to Cart