ENA371463510 Enamine

3-methylimidazo[4,3-b][1,3]thiazole-2-carboxylic acid

  • MDL number MFCD22064468
  • Cas Number : 1368324-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109479
Availability details for
Quantity
  • Availability
  • Add to Cart