ENA371463511 Enamine

methyl 2-(3,5-dichlorophenyl)-2-hydroxyacetate

  • MDL number MFCD16693851
  • Cas Number : 1249705-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109480
Availability details for
Quantity
  • Availability
  • Add to Cart