ENA371463513 Enamine

1-ethylcyclopentane-1-carbonitrile

  • MDL number MFCD20882675
  • Cas Number : 49826-26-8
  • Molecular Weight : 123.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109482
Availability details for
Quantity
  • Availability
  • Add to Cart