ENA371463515 Enamine

1,3-bis(2-fluorophenyl)propan-2-one

  • MDL number MFCD16204277
  • Cas Number : 1425-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109485
Availability details for
Quantity
  • Availability
  • Add to Cart