ENA371463521 Enamine

5-tert-butylthiophene-2-carboxylic acid

  • MDL number MFCD01566583
  • Cas Number : 29212-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109496
Availability details for
Quantity
  • Availability
  • Add to Cart