ENA371463522 Enamine

4-(trifluoromethyl)quinoline

  • MDL number MFCD01862095
  • Cas Number : 25199-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109497

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109497
Availability details for
Quantity
  • Availability
  • Add to Cart