ENA371463535 Enamine

3-methyl-1-benzothiophene-5-carbonitrile

  • MDL number MFCD05664868
  • Cas Number : 19404-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100306
Availability details for
Quantity
  • Availability
  • Add to Cart