ENA371463536 Enamine

methyl 3-amino-4-(benzyloxy)benzoate

  • MDL number MFCD09054786
  • Cas Number : 927802-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109558

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109558
Availability details for
Quantity
  • Availability
  • Add to Cart