ENA371463537 Enamine

2,2-dimethyl-3-(oxan-4-yl)propan-1-amine

  • MDL number MFCD16855934
  • Cas Number : 1262408-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109561
Availability details for
Quantity
  • Availability
  • Add to Cart