ENA371463539 Enamine

3-[(4-chloro-3-nitrophenyl)methyl]imidazolidine-2,4-dione

  • MDL number MFCD11547887
  • Cas Number : 1097788-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109564
Availability details for
Quantity
  • Availability
  • Add to Cart