ENA371463540 Enamine

N-methoxy-N-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxamide

  • MDL number MFCD18918961
  • Cas Number : 1394041-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109565
Availability details for
Quantity
  • Availability
  • Add to Cart