ENA371463541 Enamine

1-[(4-chloro-3-nitrophenyl)methyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD11547889
  • Cas Number : 1097788-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109566
Availability details for
Quantity
  • Availability
  • Add to Cart