ENA371463542 Enamine

1-[(4-chloro-3-nitrophenyl)methyl]pyrrolidine-2,5-dione

  • MDL number MFCD11547891
  • Cas Number : 1097788-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109567
Availability details for
Quantity
  • Availability
  • Add to Cart