ENA371463543 Enamine

N-(2-cyclopropyl-2-hydroxyethyl)-2-(trifluoromethyl)benzamide

  • MDL number MFCD19129747
  • Cas Number : 1273988-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109569
Availability details for
Quantity
  • Availability
  • Add to Cart