ENA371463544 Enamine

2-(4-phenyl-1H-1,2,3-triazol-1-yl)acetic acid

  • MDL number MFCD02927730
  • Cas Number : 51720-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107888

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-107888
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart