ENA371463548 Enamine

2-hydroxy-5-[(4-methoxypiperidine-1-carbonyl)amino]benzoic acid

  • MDL number MFCD22378603
  • Cas Number : 1307075-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107921
Availability details for
Quantity
  • Availability
  • Add to Cart