ENA371463554 Enamine

tert-butyl 5-amino-3-methyl-1H-pyrazole-1-carboxylate

  • MDL number MFCD11656647
  • Cas Number : 1065204-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109665

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109665
Availability details for
Quantity
  • Availability
  • Add to Cart