ENA371463563 Enamine

1-[1-(2-methanesulfonylethyl)-3,5-dimethyl-1H-pyrazol-4-yl]ethan-1-one

  • MDL number MFCD16861498
  • Cas Number : 1344063-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109682
Availability details for
Quantity
  • Availability
  • Add to Cart