ENA371463566 Enamine

4-(aminomethyl)-N-ethylaniline

  • MDL number MFCD16673869
  • Cas Number : 1228098-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109687
Availability details for
Quantity
  • Availability
  • Add to Cart