ENA371463567 Enamine

1-[(4-bromo-3-fluorophenyl)methyl]piperazine

  • MDL number MFCD16769813
  • Cas Number : 1249436-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109688
Availability details for
Quantity
  • Availability
  • Add to Cart