ENA371463569 Enamine

2-(3,5-dichlorophenyl)-2-(dimethylamino)acetic acid

  • MDL number MFCD12149928
  • Cas Number : 1218320-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109690
Availability details for
Quantity
  • Availability
  • Add to Cart