ENA371463570 Enamine

N-cyclopropyl-3,5-difluoro-4-hydrazinylbenzamide

  • MDL number MFCD18285494
  • Cas Number : 1343718-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109691
Availability details for
Quantity
  • Availability
  • Add to Cart