ENA371463573 Enamine

2-[2-(3-aminophenyl)-1H-imidazol-1-yl]acetic acid

  • MDL number MFCD09908296
  • Cas Number : 1247935-00-7
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(3-aminophenyl)-1H-imidazol-1-yl]acetic acid
Inchi Code 1S/C11H11N3O2/c12-9-3-1-2-8(6-9)11-13-4-5-14(11)7-10(15)16/h1-6H,7,12H2,(H,15,16)
Inchi Key ZMCIUPRLOYUAOS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109697
Availability details for
Quantity
  • Availability
  • Add to Cart