ENA371463588 Enamine

5-bromo-7-fluoro-2,3-dihydro-1H-indole

  • MDL number MFCD09739495
  • Cas Number : 954258-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109731
Availability details for
Quantity
  • Availability
  • Add to Cart